Palladium 10% on activated carbon powder wetted with ca. 50% water for use as a catalyst, unreduced

Supplier: Thermo Scientific Chemicals

38305-06 38305-14 38305-22 38305-22 38305-06 38305-14
CAAA38305-14EA 1306.66 CAD
AA38305-06 AA38305-22 CAAA38305-14 CAAA38305-22 CAAA38305-06 AA38305-14
Palladium 10% on activated carbon powder wetted with ca. 50% water for use as a catalyst, unreduced
Palladium

Sold on a dry weight basis. Unit weight excludes water weight


Physical Form: 900-1100 m²/g

Formula: Pd
MW: 106.420 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00011167
CAS Number: 7440-05-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR