Palladium 5% on activated carbon paste for use as a catalyst A102023-5
Supplier: Thermo Scientific Chemicals
38300-06
38300-14
38300-22
CAAA38300-06EA
130.67
CAD
CAAA38300-06
CAAA38300-14
CAAA38300-22
Palladium 5% on activated carbon paste for use as a catalyst A102023-5
Palladium
Appearance: Black
Odor: Odorless
Ordering information: Sold on a dry weight basis. Unit weight excludes water weight. Nominally 50% water wet
Formula:
Pd MW: 106.420 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD03457879 CAS Number: 7440-05-3 EINECS: 231-115-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...