Palladium 5% on activated carbon powder wetted with ca. 50% water, eggshell, unreduced Type A401102-5
Supplier: Thermo Scientific Chemicals
38299-06
38299-14
38299-22
CAAA38299-06EA
130.67
CAD
CAAA38299-06
CAAA38299-14
CAAA38299-22
Palladium 5% on activated carbon powder wetted with ca. 50% water, eggshell, unreduced Type A401102-5
Palladium
Applications: Aromatic nitro group hydrogenation
CAS No.: 7440-05-3
Physical Form: 900-1100m2/g
Appearance: Black
Odor: Odorless
MDL No.: MFCD03457879
Applications: Aromatic nitro group hydrogenation
Formula:
Pd Storage Temperature: Ambient |
MDL Number:
MFCD00011167 CAS Number: 7440-05-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...