Palladium 3% on activated carbon wetted with ca. 50% water, powder hydrogenation catalyst, reduced
Supplier: Thermo Scientific Chemicals
38296-06
38296-14
38296-22
CAAA38296-06EA
98.91
CAD
CAAA38296-06
CAAA38296-14
CAAA38296-22
Palladium 3% on activated carbon wetted with ca. 50% water, powder hydrogenation catalyst, reduced
Palladium
Sold on a dry weight basis. Unit weight excludes water weight
CAS No.: 7440-05-3
Physical Form: 900-1100m2/g
Odor: Odorless
MDL No.: MFCD03457879
Formula:
Pd Storage Temperature: Ambient |
MDL Number:
MFCD00011167 CAS Number: 7440-05-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...