Semiquantitative Standard 2, Specpure®, Thermo Scientific
Supplier: Thermo Scientific Chemicals
36770-AE
36770-AP
CAAA36770-AEEA
748.92
CAD
CAAA36770-AE
CAAA36770-AP
Semiquantitative Standard 2, Specpure®, Thermo Scientific
Standards
Cationic Standards
Multi-Element Standards
Certificate of Analysis included
Molecular Formula: Matrix: 40% Aqua Regia/tr. HF
Physical Form: Liquid
UN No.: 3093
MDL No.: MFCD02091743
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...