Bis[2-carboxyethylgermanium(IV)] sesquioxide ≥99.7%

Supplier: Thermo Scientific Chemicals
Warning

36758-06 36758-14 36758-22
CAAA36758-06EA 116.76 CAD
CAAA36758-06 CAAA36758-14 CAAA36758-22
Bis[2-carboxyethylgermanium(IV)] sesquioxide ≥99.7%
Bis[2-carboxyethylgermanium(IV)] sesquioxide

Purity: 99.7% CAS No.: 12758-40-6 Molecular Formula: C6H10Ge2O7 Formula Weight: 339.25 Physical Form: Powder Odor: Odorless Melting Point: 320° dec. MDL No.: MFCD00049319

Formula: C₆H₁₀Ge₂O₇
Melting Pt: 320 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00049319
CAS Number: 12758-40-6
EINECS: 235-800-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR