TCLP Standard Solution 1, Specpure®, Thermo Scientific

Supplier: Thermo Scientific Chemicals

36743-AE 36743-AP
CAAA36743-AEEA 229.1 CAD
CAAA36743-AE CAAA36743-AP
TCLP Standard Solution 1, Specpure®, Thermo Scientific
Standards Cationic Standards Multi-Element Standards

Molecular Formula: Matrix: 2% HNO3
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD02091705

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR