Calcium phosphate tribasic (34 - 40% Ca)

Supplier: Thermo Scientific Chemicals
Warning

36731-36 36731-A3 36731-A9
CAAA36731-36EA 70.47 CAD
CAAA36731-36 CAAA36731-A3 CAAA36731-A9
Calcium phosphate tribasic (34 - 40% Ca)
Calcium phosphate tribasic

CAS No.: 12167-74-7 Molecular Formula: Ca10(OH)2(PO4)6 (approximate) Formula Weight: 1004.67 Physical Form: Powder Odor: Odorless MDL No.: MFCD00010904

Formula: Ca₅HO₁₃P₃
Storage Temperature: Ambient
MDL Number: MFCD00010904
CAS Number: 12167-74-7
EINECS: 235-330-6

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR