Lead(II)-2-ethylhexanoate

Supplier: Thermo Scientific Chemicals
Danger

36711-18 36711-30
CAAA36711-18EA 96.54 CAD
CAAA36711-18 CAAA36711-30
Lead(II)-2-ethylhexanoate
Lead(II)-2-ethylhexanoate

CAS No.: 301-08-6 Molecular Formula: C16H30O4Pb Formula Weight: 493.61 Physical Form: Viscous Liquid Density: 1.56 Odor: Odorless UN No.: 3082 MDL No.: MFCD00014003

Formula: C₁₆H₃₀O₄Pb
Density: 1.56
Storage Temperature: Ambient
MDL Number: MFCD00014003
CAS Number: 301-08-6
EINECS: 206-107-0
UN: 3082
ADR: 9,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR