Bismuth(III) 2-ethylhexanoate

Supplier: Thermo Scientific Chemicals
Warning

36710-06 36710-14 36710-22
CAAA36710-06EA 55.62 CAD
CAAA36710-06 CAAA36710-14 CAAA36710-22
Bismuth(III) 2-ethylhexanoate
Bismuth (III) 2-ethylhexanoate

Contains 2-ethylhexanoic acid


CAS No.: 67874-71-9 Molecular Formula: C24H45BiO6 Formula Weight: 638.61 Physical Form: Viscous liquid MDL No.: MFCD00043803

Formula: C₂₄H₄₅BiO₆
Storage Temperature: Ambient
MDL Number: MFCD00043803
CAS Number: 67874-71-9
EINECS: 267-499-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR