Iron nickel ≥99.5% (metals basis), powder -325 mesh
Supplier: Thermo Scientific Chemicals
|
Danger
|
36683-22
36683-36
CAAA36683-22EA
142.04
CAD
CAAA36683-22
CAAA36683-36
Iron nickel ≥99.5% (metals basis), powder -325 mesh
Iron nickel
Applications: Low expansion alloy for precision instruments, measuring tapes, weights, etc
Purity: 99.5+%
Molecular Formula: Ni:Fe; 50:50 wt%
Physical Form: Powder
Appearance: Grey
Odor: Odorless
UN No.: 3089
MDL No.: MFCD01741928
Applications: Low expansion alloy for precision instruments, measuring tapes, weights, etc
Formula:
Ni:Fe;₅₀:₅₀% Storage Temperature: Ambient |
MDL Number:
MFCD01741928 UN: 3089 ADR: 4.1,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...