Yttrium(III) 2-ethylhexanoate ≥99.8% (metals basis)
Supplier: Thermo Scientific Chemicals
|
Warning
|
36661-14
36661-22
CAAA36661-06EA
129.18
CAD
CAAA36661-06
CAAA36661-14
CAAA36661-22
Yttrium(III) 2-ethylhexanoate ≥99.8% (metals basis)
Yttrium (III) 2-ethylhexanoate
Purity: 99.8%
CAS No.: 103470-68-4
Molecular Formula: C24H45O6Y
Formula Weight: 518.16
Physical Form: Powder
Formula:
C₂₄H₄₅O₆Y Storage Temperature: Ambient |
MDL Number:
MFCD00070441 CAS Number: 103470-68-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...