Erbium(III) isopropoxide
Supplier: Thermo Scientific Chemicals
|
Danger
|
36613-03
36613-06
CAAA36613-03EA
118.66
CAD
CAAA36613-03
CAAA36613-06
Erbium(III) isopropoxide
Erbium (III) isopropoxide
CAS No.: 14814-07-4
Molecular Formula: C9H21ErO3
Formula Weight: 344.52
Flash Point: 58°(138°F)
Physical Form: Powder
Sensitivity: Air and Moisture Sensitive
UN No.: 2925
MDL No.: MFCD00144253
Formula:
C₉H₂₁ErO₃ Melting Pt: 325 °C (dec.) Flash Pt: 58 °C (138 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00144253 CAS Number: 14814-07-4 UN: 2925 ADR: 4.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...