Aluminum calcium isopropoxide 10% (w/v) in isopropanol (IPA) , 99.8% (metals basis)
Supplier: Thermo Scientific Chemicals
|
Danger
|
36594-AC
36594-AE
CAAA36594-ACEA
145.36
CAD
CAAA36594-AC
CAAA36594-AE
Aluminum calcium isopropoxide 10% (w/v) in isopropanol (IPA) , 99.8% (metals basis)
Aluminum calcium isopropoxide
Purity: 99.8%
CAS No.: 23275-27-6
Molecular Formula: Ca[Al(OCH(CH3)2)4]2
Formula Weight: 566.30
Physical Form: Liquid
Sensitivity: Moisture Sensitive
Odor: Alcohol-like
Boiling Point: 230-240°/4.0mm
UN No.: 1219
MDL No.: MFCD00143586
Formula:
Ca[Al(OCH(CH₃)₂)₄]₂ Boiling Pt: 230…240 °C (4.0 mmHg) Storage Temperature: Ambient |
MDL Number:
MFCD00143586 CAS Number: 23275-27-6 UN: 1219 ADR: 3,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...