Antimony(III) isopropoxide ≥99.9% (metals basis)
Supplier: Thermo Scientific Chemicals
|
Warning
|
36589-06
36589-14
CAAA36589-06EA
165.9
CAD
CAAA36589-06
CAAA36589-14
Antimony(III) isopropoxide ≥99.9% (metals basis)
Antimony (III) isopropoxide
Purity: 99.9%
CAS No.: 18770-47-3
Molecular Formula: Sb[OCH(CH3)2]3
Formula Weight: 298.80
Flash Point: 77°(141°F)
Physical Form: Liquid
Density: 1.31
Sensitivity: Moisture Sensitive
Appearance: Colorless
Boiling Point: 68°/3mm
UN No.: 3082
MDL No.: MFCD00143623
Formula:
C₉H₂₁O₃Sb MW: 298.80 g/mol Boiling Pt: 68 °C (3 mmHg) Density: 1.31 Flash Pt: 77 °C (141 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00143623 CAS Number: 18770-47-3 EINECS: 242-561-6 UN: 3082 ADR: 9 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...