Barium ethoxide 10% w/v ethanol , 99.5% Metals basis

Supplier: Thermo Scientific Chemicals
Danger

36588-AC 36588-AE
CAAA36588-ACEA 133.67 CAD
CAAA36588-AC CAAA36588-AE
Barium ethoxide 10% w/v ethanol , 99.5% Metals basis
Barium ethoxide

Purity: 99.5% CAS No.: 2914-19-4 Molecular Formula: C4H10BaO2 Formula Weight: 227.42 Physical Form: Liquid Sensitivity: Moisture Sensitive Appearance: Yellow UN No.: 3274 MDL No.: MFCD00050488

Formula: C₄H₁₀BaO₂
MW: 227.449 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00050488
CAS Number: 2914-19-4
EINECS: 220-829-3
UN: 3274
ADR: 3 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR