Gallium(III) isopropoxide mixture of oligomers ≥99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
36583-03
36583-06
CAAA36583-03EA
167.48
CAD
CAAA36583-03
CAAA36583-06
Gallium(III) isopropoxide mixture of oligomers ≥99%
Gallium(III) isopropoxide
Purity: 99%
CAS No.: 4452-61-3
Molecular Formula: C9H21GaO3
Formula Weight: 246.98
Flash Point: 68°(154°F)
Physical Form: Liquid
Density: 1.35
Sensitivity: Moisture Sensitive
Appearance: Yellow
Melting Point: 72°
Boiling Point: dec.
UN No.: 1993
MDL No.: MFCD00144389
Formula:
C₉H₂₁GaO₃ Boiling Pt: 120 °C (1 mmHg) Density: 1.35 Flash Pt: 68 °C (154 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00144389 CAS Number: 4452-61-3 UN: 1993 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...