Neodymium(III) isopropoxide 2,5% (w/v) in 2-propanol/toluene , 99% (REO, rare earth oxide basis)

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Neodymium(III) i-propoxide

36573-AC 36573-AE
CAAA36573-ACEA 189.6 CAD
CAAA36573-AC CAAA36573-AE
Neodymium(III) isopropoxide 2,5% (w/v) in 2-propanol/toluene , 99% (REO, rare earth oxide basis)
Neodymium(III) isopropoxide

Purity: 99% CAS No.: 19236-15-8 Molecular Formula: C9H21NdO3 Formula Weight: 321.30 Physical Form: Liquid Sensitivity: Moisture Sensitive Appearance: Blue UN No.: 1993 MDL No.: MFCD00058819

Formula: C₉H₂₁NdO₃
MW: 321.503 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00058819
CAS Number: 19236-15-8
UN: 1993
ADR: 3 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR