Niobium isopropoxide 10% (w/v) in isopropanol/hexane (50:50) , 99% (metals basis)

Supplier: Thermo Scientific Chemicals
Danger

36572-AC 36572-AE
CAAA36572-ACEA 233.84 CAD
CAAA36572-AC CAAA36572-AE
Niobium isopropoxide 10% (w/v) in isopropanol/hexane (50:50) , 99% (metals basis)
Niobium isopropoxide

Purity: 99% CAS No.: 18368-80-4 Molecular Formula: C15H35NbO5 Formula Weight: 388.35 Physical Form: Liquid Sensitivity: Moisture Sensitive UN No.: 1993 MDL No.: MFCD00144915

Formula: C₁₅H₃₅NbO₅
Storage Temperature: Ambient
MDL Number: MFCD00144915
CAS Number: 18368-80-4
UN: 1993
ADR: 3,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR