Niobium-n-butoxide ≥99% (metals basis)
Supplier: Thermo Scientific Chemicals
|
Warning
|
36570-03
36570-06
36570-14
CAAA36570-03EA
91.48
CAD
CAAA36570-03
CAAA36570-06
CAAA36570-14
Niobium-n-butoxide ≥99% (metals basis)
Niobium-n-butoxide
Purity: 99%
CAS No.: 51030-47-8
Molecular Formula: C20H45NbO5
Formula Weight: 458.12
Physical Form: Liquid
Density: 1.09
Sensitivity: Moisture Sensitive
Boiling Point: 197°/0.15mm
MDL No.: MFCD00144916
Formula:
C₂₀H₄₅NbO₅ MW: 458.12 g/mol Boiling Pt: 197 °C (0.15 mmHg) Density: 1,09 Storage Temperature: Ambient |
MDL Number:
MFCD00144916 CAS Number: 51030-47-8 EINECS: 256-923-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...