Tin(IV) isopropoxide 10% (w/v) in 2-propanol/toluene , 98% (metals basis)
Supplier: Thermo Scientific Chemicals
|
Danger
|
36563-AA
36563-AC
36563-AE
36563-AP
CAAA36563-AAEA
63.67
CAD
CAAA36563-AA
CAAA36563-AC
CAAA36563-AE
CAAA36563-AP
Tin(IV) isopropoxide 10% (w/v) in 2-propanol/toluene , 98% (metals basis)
Tin(IV) isopropoxide
Purity: 98%
CAS No.: 1184-61-8
Molecular Formula: Sn[OCH(CH3)2]4
Formula Weight: 355.06
Physical Form: Liquid
Sensitivity: Moisture Sensitive
UN No.: 1993
MDL No.: MFCD00145408
Formula:
C₁₂H₂₈O₄Sn Storage Temperature: Ambient |
MDL Number:
MFCD00145408 CAS Number: 1184-61-8 UN: 1993 ADR: 3,II |
Specification Test Results
Form | Liquid |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...