Tris(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')lanthanum(III) 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: La(TMHD)3, Tris(2,2,6,6-tetramethyl-3,5-heptanedionato)lanthanum(III)

36465-03 36465-06
CAAA36465-03EA 127.35 CAD
CAAA36465-03 CAAA36465-06
Tris(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')lanthanum(III) 98%
Tris(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')lanthanum(III)

Superconductor MOCVD Precursor Subl. 210°/0.2mm


Purity: 98% CAS No.: 14319-13-2 Molecular Formula: C33H57LaO6 Formula Weight: 688.72 Physical Form: Powder Sensitivity: Hygroscopic Boiling Point: 370° dec. MDL No.: MFCD00010466

Formula: C₃₃H₅₇LaO₆
MW: 688.71 g/mol
Boiling Pt: 370 °C (dec.)
Melting Pt: 240 °C
Storage Temperature: Ambient
MDL Number: MFCD00010466
CAS Number: 14319-13-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR