Nickel copper iron manganese (Ni:Cu:Fe:Mn; 63:28-34:2.5 max:2 max wt%), rods, Ø 1.27cm (0.5 in)
Supplier: Thermo Scientific Chemicals
|
Warning
|
36457-BS
36457-BW
CAAA36457-BSEA
195.92
CAD
CAAA36457-BS
CAAA36457-BW
Nickel copper iron manganese (Ni:Cu:Fe:Mn; 63:28-34:2.5 max:2 max wt%), rods, Ø 1.27cm (0.5 in)
Nickel copper iron manganese
Molecular Formula: Ni:Cu:Fe:Mn; 63:28-34:2.5 max:2 max wt%
Physical Form: ≈104g/10cm, cold drawn stress relieved, ASTM B164
MDL No.: MFCD02091727
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...