Gold nickel ≥99.85% (metals basis), wire, annealed, Ø 0.5 mm (0.02 in)
Supplier: Thermo Scientific Chemicals
|
Warning
|
36451-BS
36451-BW
36451-G1
CAAA36451-BSEA
206.98
CAD
CAAA36451-BS
CAAA36451-BW
CAAA36451-G1
Gold nickel ≥99.85% (metals basis), wire, annealed, Ø 0.5 mm (0.02 in)
Gold nickel
Purity: 99.85%
Molecular Formula: Au:Ni; 82:18 wt%
Physical Form: ≈0.32g/10cm, Temper: soft
MDL No.: MFCD02091707
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...