Phenylazoformic acid 2-phenylhydrazide compound with 1,5-diphenylcarbohydrazide ACS
Supplier: Thermo Scientific Chemicals
|
Warning
|
36442-09
36442-18
CAAA36442-09EA
82.63
CAD
CAAA36442-09
CAAA36442-18
Phenylazoformic acid 2-phenylhydrazide compound with 1,5-diphenylcarbohydrazide ACS
Phenylazoformic acid 2-phenylhydrazide compound with 1,5-diphenylcarbohydrazide
CAS #: [538-62-5] with [140-22-7]
CAS No.: 10329-15-4
Molecular Formula: C6H5N=NCONHNHC6H5.C6H5NHNHCONHNHC6H5
Formula Weight: 482.55
Physical Form: Powder
Appearance: Orange
Odor: Odorless
Melting Point: 154-156°
Beilstein Reference: 6110063
MDL No.: MFCD00013218
Formula:
C₂₆H₂₆N₈O₂ Melting Pt: 154…156 °C Storage Temperature: Ambient |
MDL Number:
MFCD00013218 CAS Number: 10329-15-4 Merck Index: 14,3322 |
Specification Test Results
Residue after ignition | Pass test |
Solubility in acetone | Pass test. |
Sensitivity | Pass test. |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...