D-(-)-Isoascorbic acid ≥98% (by titrimetric analysis)

Supplier: Thermo Scientific Chemicals

36366-22 36366-36
CAAA36366-22EA 71.8 CAD
CAAA36366-22 CAAA36366-36
D-(-)-Isoascorbic acid ≥98% (by titrimetric analysis)
D(-)-Isoascorbic acid

Purity: 99% CAS No.: 89-65-6 Molecular Formula: C6H8O6 Formula Weight: 176.12 Physical Form: Powder Odor: Odorless Merck Reference: 14,5126 MDL No.: MFCD00005378

Formula: C₆H₈O₆
Melting Pt: 169…172 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00005378
CAS Number: 89-65-6
EINECS: 201-928-0
Merck Index: 14,5126

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR