Indium tin oxide ≥99.99% (metals basis)
Supplier: Thermo Scientific Chemicals
|
Warning
|
36348-06
36348-14
36348-22
CAAA36348-06EA
66.83
CAD
CAAA36348-06
CAAA36348-14
CAAA36348-22
Indium tin oxide ≥99.99% (metals basis)
Indium tin oxide
Purity: 99.99%
CAS No.: 50926-11-9
Molecular Formula: In2O3:SnO2; 90:10 wt%
Physical Form: -325+500 Mesh Powder
Odor: Odorless
MDL No.: MFCD00171662
Formula:
InOSn Storage Temperature: Ambient |
MDL Number:
MFCD00171662 CAS Number: 50926-11-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...