Zirconium disilicide ≥99.5% (metals basis excluding Hf) (max. 4% Hf)
Supplier: Thermo Scientific Chemicals
Synonyms:
Zirconium silicide
36322-14
36322-22
CAAA36322-14EA
176.96
CAD
CAAA36322-14
CAAA36322-22
Zirconium disilicide ≥99.5% (metals basis excluding Hf) (max. 4% Hf)
Zirconium disilicide
Purity: 99.5%
CAS No.: 12039-90-6
Molecular Formula: ZrSi2 (may contain other phases)
Formula Weight: 147.39
Physical Form: -325 Mesh Powder
Density: 4.88
Appearance: Grey
Odor: Odorless
MDL No.: MFCD00151547
Formula:
Si₂Zr MW: 147.39 g/mol Density: 4.88 Storage Temperature: Ambient |
MDL Number:
MFCD00151547 CAS Number: 12039-90-6 EINECS: 234-911-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...