2-Hydroxy-2-propanesulfonic acid monosodium salt ≥97%
Supplier: Thermo Scientific Chemicals
36256-22
36256-36
CAAA36256-22EA
144.41
CAD
CAAA36256-22
CAAA36256-36
2-Hydroxy-2-propanesulfonic acid monosodium salt ≥97%
2-Hydroxy-2-propanesulfonic acid monosodium salt
Purity: 97%
CAS No.: 540-92-1
Molecular Formula: C3H7NaO4S
Formula Weight: 162.14
Physical Form: Crystalline
MDL No.: MFCD00031395
Formula:
C₃H₇NaO₄S MW: 162.14 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00031395 CAS Number: 540-92-1 EINECS: 208-761-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...