Yttrium aluminum oxide ≥99.99% (metals basis)

Supplier: Thermo Scientific Chemicals

36241-14 36241-22
CAAA36241-14EA 194.34 CAD
CAAA36241-14 CAAA36241-22
Yttrium aluminum oxide ≥99.99% (metals basis)
Yttrium aluminum oxide

Applications: Can be used as a sputtering target to prepare bubble memory devices.


Purity: 99.99% CAS No.: 12005-21-9 Molecular Formula: Y3Al5O12 Formula Weight: 593.62 Physical Form: 3-12mm Fused Lump Density: 4.65 Refractive Index: 1.833 Melting Point: 1760° MDL No.: MFCD00213958


Applications: Can be used as a sputtering target to prepare bubble memory devices.

Formula: Y₃Al₅O₁₂
MW: 593.62 g/mol
Melting Pt: 1760 °C
Density: 4.65
Storage Temperature: Ambient
MDL Number: MFCD00213958
CAS Number: 12005-21-9
EINECS: 234-465-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR