D-Lactose monohydrate ACS
Supplier: Thermo Scientific Chemicals
36218-36
36218-A3
CAAA36218-36EA
105.42
CAD
CAAA36218-36
CAAA36218-A3
D-Lactose monohydrate ACS
D-Lactose monohydrate
CAS No.: 64044-51-5
Molecular Formula: C12H22O11.H2O
Formula Weight: 360.32 (342.30anhy)
Physical Form: Crystalline
Density: 1.525
Melting Point: ca 218° dec.
Beilstein Reference: 3768231
MDL No.: MFCD00150747
Formula:
C₁₂H₂₂O₁₁·1H₂O Melting Pt: ∼ 218 °C (dec.) Density: 1.525 Storage Temperature: Ambient |
MDL Number:
MFCD00150747 CAS Number: 64044-51-5 EINECS: 200-559-2 Merck Index: 14,5343 |
Specification Test Results
Insoluble matter | Pass test |
Residue after ignition | Pass test. |
Dextrose | Pass test. |
Sucrose | Pass test. |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...