Bis(cyclopentadienyl)hafnium dichloride
Supplier: Thermo Scientific Chemicals
|
Danger
|
36105-03
36105-06
36105-14
CAAA36105-03EA
99.07
CAD
CAAA36105-03
CAAA36105-06
CAAA36105-14
Bis(cyclopentadienyl)hafnium dichloride
Bis(cyclopentadienyl)hafnium dichloride
CAS No.: 12116-66-4
Molecular Formula: C10H10Cl2Hf
Formula Weight: 379.59
Physical Form: Crystalline, Packaged under argon
Sensitivity: Air and Moisture Sensitive
Melting Point: 230-233°
UN No.: 3261
MDL No.: MFCD00001438
Formula:
C₁₀H₁₀Cl₂Hf Melting Pt: 230…233 °C Storage Temperature: Ambient |
MDL Number:
MFCD00001438 CAS Number: 12116-66-4 EINECS: 235-177-5 UN: 3261 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...