Cerium disilicide ≥99.9% (REO, rare earth oxide basis)
Supplier: Thermo Scientific Chemicals
35836-06
35836-14
CAAA35836-06EA
260.7
CAD
CAAA35836-06
CAAA35836-14
Cerium disilicide ≥99.9% (REO, rare earth oxide basis)
Cerium disilicide
Purity: 99.9%
CAS No.: 12014-85-6
Molecular Formula: CeSi2
Formula Weight: 196.29
Physical Form: 3-15mm Pieces
Density: 5.31
Appearance: Grey
Odor: Odorless
Melting Point: 1620°
MDL No.: MFCD00168082
Formula:
CeSi₂ Melting Pt: 1620 °C Density: 5,31 Storage Temperature: Ambient |
MDL Number:
MFCD00168082 CAS Number: 12014-85-6 EINECS: 234-602-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...