Copper(II) isopropoxide ≥98% (metals basis)
Supplier: Thermo Scientific Chemicals
|
Warning
|
35821-06
35821-14
CAAA35821-06EA
347.6
CAD
CAAA35821-06
CAAA35821-14
Copper(II) isopropoxide ≥98% (metals basis)
Copper (II) isopropoxide
Purity: 98%
CAS No.: 23578-23-6
Molecular Formula: [(CH3)2CHO]2Cu
Formula Weight: 181.73
Physical Form: Powder
Sensitivity: Moisture Sensitive
UN No.: 3180
MDL No.: MFCD00074982
Formula:
C₆H₁₄CuO₂ MW: 181.73 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00074982 CAS Number: 23578-23-6 UN: 3181 ADR: 4.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...