Zirconium carbide ≥99.5% (metals basis excluding Hf) (max. 200 ppm Hf)
Supplier: Thermo Scientific Chemicals
|
Warning
|
35808-18
35808-30
CAAA35808-18EA
145.52
CAD
CAAA35808-18
CAAA35808-30
Zirconium carbide ≥99.5% (metals basis excluding Hf) (max. 200 ppm Hf)
Zirconium carbide
Purity: 99.5%
CAS No.: 12070-14-3
Molecular Formula: ZrC
Formula Weight: 103.23
Physical Form: -325 Mesh Powder
Density: 6.73
Appearance: Grey
Odor: Odorless
Melting Point: 3540°
UN No.: 3178
MDL No.: MFCD00049640
Formula:
CZr Melting Pt: 3540 °C Density: 6.73 Storage Temperature: Ambient |
MDL Number:
MFCD00049640 CAS Number: 12070-14-3 EINECS: 235-125-1 UN: 3178 ADR: 4.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...