Calcium wolframate ≥99.78% (metals basis)

Supplier: Thermo Scientific Chemicals

35669-18 35669-30
CAAA35669-18EA 156.26 CAD
CAAA35669-18 CAAA35669-30
Calcium wolframate ≥99.78% (metals basis)
Calcium wolframate

Purity: 99.78% CAS No.: 7790-75-2 Molecular Formula: CaWO4 Formula Weight: 287.93 Physical Form: -325 Mesh Powder Melting Point: 1620° MDL No.: MFCD00010913

Formula: CaO₄W
Melting Pt: 1620 °C
Storage Temperature: Ambient
MDL Number: MFCD00010913
CAS Number: 7790-75-2
EINECS: 232-219-4
Merck Index: 14,1712

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR