Potassium hydrogen phthalate standardised solution 0.05 N in aqueous solution
Supplier: Thermo Scientific Chemicals
Synonyms:
Potassium phthalate monobasic, Potassium biphthalate, Potassium Acid Phthalate, KHP, Phthalic Acid Monopotassium Salt, Phthalic acid potassium salt
35603-AP
35603-K2
CAAA35603-APEA
57.83
CAD
CAAA35603-AP
CAAA35603-K2
Potassium hydrogen phthalate standardised solution 0.05 N in aqueous solution
Potassium hydrogen phthalate
CAS No.: 877-24-7
Molecular Formula: C8H5KO4
Formula Weight: 204.23
Physical Form: Liquid
Odor: Odorless
MDL No.: MFCD00013070
Formula:
HOOCC₆H₄COOK MW: 204.22 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00013070 CAS Number: 877-24-7 EINECS: 212-889-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...