Potassium iodate standardised solution 0.1 N in aqueous solution

Supplier: Thermo Scientific Chemicals

35586-K2 35586-K7
CAAA35586-K2EA 122.77 CAD
CAAA35586-K2 CAAA35586-K7
Potassium iodate standardised solution 0.1 N in aqueous solution
Potassium iodate

CAS No.: 7758-05-6 Molecular Formula: KIO3 Formula Weight: 214.00 Physical Form: Liquid Appearance: Colorless Odor: Odorless MDL No.: MFCD00011406

Formula: KlO₃
Storage Temperature: Ambient
MDL Number: MFCD00011406
CAS Number: 7758-05-6
EINECS: 231-831-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR