Ammonium dimolybdate 4% (w/v) in aqueous solution

Supplier: Thermo Scientific Chemicals

35578-AP 35578-K2
CAAA35578-APEA 118.18 CAD
CAAA35578-AP CAAA35578-K2
Ammonium dimolybdate 4% (w/v) in aqueous solution
Ammonium dimolybdate

Molecular Formula: (NH4)2Mo2O7 Formula Weight: 339.95 Physical Form: Liquid MDL No.: MFCD00010882

Formula: H₈Mo₂N₂O₇
Storage Temperature: Ambient
MDL Number: MFCD00010882
CAS Number: 27546-07-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR