Ammonia solution 50% v/v aqueous solution

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Ammonia aqueous, Ammonium hydroxide solution, Ammonium hydroxide, Ammonia water

35574-AP 35574-K2 35574-K7
CAAA35574-APEA 103.02 CAD
CAAA35574-AP CAAA35574-K2 CAAA35574-K7
Ammonia solution 50% v/v aqueous solution
Ammonia solution

CAS No.: 1336-21-6 Molecular Formula: H5NO Formula Weight: 35.05 Physical Form: Liquid Density: 0.94 Appearance: Colorless UN No.: 2672 MDL No.: MFCD00066650

Formula: NH₃
MW: 35.05 g/mol
Density: 0.94
Storage Temperature: Ambient
MDL Number: MFCD00066650
CAS Number: 1336-21-6
EINECS: 215-647-6
UN: 2672
ADR: 8 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR