Bromide, Ion chromatography Standard Solution, Specpure®, Br- 1000 µg/ml, Thermo Scientific
Supplier: Thermo Scientific Chemicals
35552-AP
35552-AE
CAAAA35552-APEA
385.52
CAD
CAAAA35552-AP
CAAA35552-AE
Bromide, Ion chromatography Standard Solution, Specpure®, Br- 1000 µg/ml, Thermo Scientific
Standards
Anionic Standards
Bromide Standards
Molecular Formula: KBr in H2O
Physical Form: Liquid
MDL No.: MFCD00134172
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...