Specpure® Nitrate, Ion chromatography standard solution, NOO₃⁻ 1000 µg/ml, Thermo Scientific
Supplier: Thermo Scientific Chemicals
35549-AP
35549-AE
CAAAA35549-APEA
385.52
CAD
CAAAA35549-AP
CAAA35549-AE
Specpure® Nitrate, Ion chromatography standard solution, NOO₃⁻ 1000 µg/ml, Thermo Scientific
Standards
Anionic Standards
Nitrate Standards
Molecular Formula: NaNO3 in H2O
Physical Form: Liquid
MDL No.: MFCD00144920
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...