Phosphate, Ion Chromatography Standard Solution, Specpure®, PO4-3 1000 µg/ml, Thermo Scientific

Supplier: Thermo Scientific Chemicals

35548-AP 35548-AE
CAAAA35548-APEA 379.2 CAD
CAAAA35548-AP CAAA35548-AE
Phosphate, Ion Chromatography Standard Solution, Specpure®, PO4-3 1000 µg/ml, Thermo Scientific

Keep refrigerated


Molecular Formula: KH2PO4 in H2O
Physical Form: Liquid
Odor: Odorless
MDL No.: MFCD00145108

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR