Sulfate, Ion Chromatography Standard Solution, Specpure®, SO₄⁻² 1000 µg/ml, Thermo Scientific
Supplier: Thermo Scientific Chemicals
35547-AP
35547-AE
CAAAA35547-APEA
385.52
CAD
CAAAA35547-AP
CAAA35547-AE
Sulfate, Ion Chromatography Standard Solution, Specpure®, SO₄⁻² 1000 µg/ml, Thermo Scientific
Standards
Anionic Standards
Sulfate standards
Molecular Formula: K2SO4 in H2O
Physical Form: Liquid
MDL No.: MFCD00145343
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...