Lithium tetraphenylborate tris(1,2-dimethoxyethane) adduct
Supplier: Thermo Scientific Chemicals
|
Warning
|
35445-02
35445-04
CAAA35445-02EA
65.73
CAD
CAAA35445-02
CAAA35445-04
Lithium tetraphenylborate tris(1,2-dimethoxyethane) adduct
Lithium tetraphenylborate tris(1,2-dimethoxyethane) adduct
CAS No.: 75965-35-4
Molecular Formula: C24H20BLi.3C4H10O2
Formula Weight: 595.54 (326.18anhy)
Physical Form: Powder
Sensitivity: Moisture Sensitive
Appearance: White to off-white
UN No.: 1325
MDL No.: MFCD00013311
Formula:
C₂₄H₂₀BLi.3C₄H₁₀O₂ Storage Temperature: Ambient |
MDL Number:
MFCD00013311 CAS Number: 75965-35-4 UN: 1325 ADR: 4.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...