Tris(6,6,7,7,8,8,8-heptafluoro-2,2-dimethyl-3,5-octanedionato-O,O')europium(III)
Supplier: Thermo Scientific Chemicals
33541-03
33541-06
33541-MD
CAAA33541-03EA
100.96
CAD
CAAA33541-03
CAAA33541-06
CAAA33541-MD
Tris(6,6,7,7,8,8,8-heptafluoro-2,2-dimethyl-3,5-octanedionato-O,O')europium(III)
Tris(6,6,7,7,8,8,8-heptafluoro-2,2-dimethyl-3,5-octanedionato-O,O')europium(III)
Fieser: 12,559 15,355 19,145 20,164
CAS No.: 17631-68-4
Molecular Formula: C30H30F21O6Eu
Formula Weight: 1037.49
Physical Form: Powder
Sensitivity: Hygroscopic
Melting Point: 203-207°
MDL No.: MFCD00064655
Formula:
C₃₀H₃₀EuF₂₁O₆ Melting Pt: 203…207 °C Storage Temperature: Ambient |
MDL Number:
MFCD00064655 CAS Number: 17631-68-4 EINECS: 241-616-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...