Manganese(II) nitrate 50% (w/w) in aqueous solution

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Manganese dinitrate, Manganous nitrate

33340-36 33340-A3
CAAA33340-36EA 96.54 CAD
CAAA33340-36 CAAA33340-A3
Manganese(II) nitrate 50% (w/w) in aqueous solution
Manganese (II) nitrate

CAS No.: 10377-66-9 Molecular Formula: MnN2O6 Formula Weight: 178.95 Physical Form: Liquid Appearance: Pink Odor: Amine-like pH: 1.0 UN No.: 3093 MDL No.: MFCD00011112

Formula: Mn(NO₃)₂
MW: 178.95 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00011112
CAS Number: 10377-66-9
EINECS: 233-828-8
UN: 3093
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR