Magnesium trisilicate

Supplier: Thermo Scientific Chemicals

33338-30 33338-A1
CAAA33338-30EA 78.05 CAD
CAAA33338-30 CAAA33338-A1
Magnesium trisilicate
Magnesium trisilicate

CAS No.: 14987-04-3 Molecular Formula: Mg2O8Si3 Formula Weight: 260.86 Physical Form: Powder Appearance: White Odor: Odorless MDL No.: MFCD00016203

Formula: Mg₂O₈Si₃
Storage Temperature: Ambient
MDL Number: MFCD00016203
CAS Number: 14987-04-3
EINECS: 239-076-7
Merck Index: 14,5687

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR