Chromium (III) sulfate hydrate, Reagent Grade
Supplier: Thermo Scientific Chemicals
|
Warning
|
33303-22
33303-36
CAAA33303-22EA
136.83
CAD
CAAA33303-22
CAAA33303-36
Chromium (III) sulfate hydrate, Reagent Grade
Chromium (III) sulphate hydrate
CAS No.: 15244-38-9
Molecular Formula: Cr2(SO4)3.xH2O
Formula Weight: 392.17(anhy)
Physical Form: Crystalline
Sensitivity: Hygroscopic
Appearance: Green to violet
Odor: Odorless
MDL No.: MFCD00016039
Formula:
Cr₂O₁₂S₃·H₂O MW: 392.17 g/mol (anhy) Storage Temperature: Ambient |
MDL Number:
MFCD00150215 CAS Number: 15244-38-9 EINECS: 233-253-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...