Silicic acid hydrate, Reagent Grade

Supplier: Thermo Scientific Chemicals
Warning

33270-22 33270-36
CAAA33270-22EA 151.68 CAD
CAAA33270-22 CAAA33270-36
Silicic acid hydrate, Reagent Grade
Silicic acid hydrate

Silicic acid hydrate is used for studying neurodegenerative diseases due to its ability to inhibit aluminum uptake. It is utilized for the manufacture catalyst and catalyst carrier. It is involved in tungsten filament production as chemical reagent and flux. Further, it is used for oil and wax decoloring.


CAS No.: 1343-98-2 Molecular Formula: SiO2.xH2O Formula Weight: 60.09(anhy) Physical Form: -100 Mesh Powder Appearance: White MDL No.: MFCD00054122

Formula: SiO₂·xH₂O
Storage Temperature: Ambient
MDL Number: MFCD00054122
CAS Number: 1343-98-2
EINECS: 215-683-2
Merck Index: 14,8490

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR